5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Formyl-4-methylphenylboronic acid pinacol ester
A912550-1G
A912550-250MG
A912550-5G
77268-866EA
210
USD
77268-866
77268-868
77268-870
5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde 98%
5-Methyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Precaución: for Research Use Only
Formula:
C₁₄H₁₉BO₃ |
MDL Number:
MFCD13181634 CAS Number: 1418129-42-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...