2-Thiophenemethanethiol 98%

Supplier: Ambeed
Danger

Synonyms: 2-(Mercaptomethyl)thiophene, Thenyl Mercaptan

A910533-1G A910533-250MG A910533-5G
77281-900EA 45.32 USD
77281-900 77281-902 77281-904
2-Thiophenemethanethiol 98%
2-Thiophenemethanethiol

Precaución: for Research Use Only

Formula: C₅H₆S₂
MW: 130.22 g/mol
MDL Number: MFCD00107135
CAS Number: 6258-63-5
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR