(4-((3-Fluorobenzyl)oxy)phenyl)methanol 97%

Supplier: Ambeed
Warning

A909896-1G A909896-5G
77268-852EA 16.48 USD
77268-852 77268-854
(4-((3-Fluorobenzyl)oxy)phenyl)methanol 97%
(4-((3-Fluorobenzyl)oxy)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₁₃FO₂
MW: 232.25 g/mol
MDL Number: MFCD03716824
CAS Number: 690969-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR