3,6-Di(4-pyridyl)-1,2,4,5-tetrazine 97%

Supplier: Ambeed
Warning

A908757-1G A908757-250MG
77281-890EA 43 USD
77281-890 77281-892
3,6-Di(4-pyridyl)-1,2,4,5-tetrazine 97%
3,6-Di(4-pyridyl)-1,2,4,5-tetrazine

Precaución: for Research Use Only

Formula: C₁₂H₈N₆
MDL Number: MFCD00129054
CAS Number: 57654-36-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR