1-Methyl-2-indoleboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: (1-Methyl-1H-indol-2-yl)boronic acid

A908509-1G A908509-5G A908509-250MG
77256-550EA 30 USD
77256-550 77256-552 77747-672
1-Methyl-2-indoleboronic acid 98%
1-Methyl-2-indoleboronic acid

Precaución: for Research Use Only

Formula: C₉H₁₀BNO₂
MW: 174.99 g/mol
MDL Number: MFCD01114668
CAS Number: 191162-40-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR