Methyl 4,4-dimethoxybutanoate 97%

Supplier: Ambeed
Warning

A908401-100MG A908401-1G A908401-250MG A908401-5G
77256-544EA 15.45 USD
77256-544 77256-546 77256-548 77595-466
Methyl 4,4-dimethoxybutanoate 97%
Methyl 4,4-dimethoxybutanoate

Precaución: for Research Use Only

Formula: C₇H₁₄O₄
MW: 162.18 g/mol
MDL Number: MFCD02683436
CAS Number: 4220-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR