3,3'-Difluorobiphenyl 98%

Supplier: Ambeed
Warning

Synonyms: 3,3'-Difluoro-1,1'-biphenyl

A908175-1G A908175-250MG A908175-5G
77281-882EA 56.65 USD
77281-882 77281-884 77281-886
3,3'-Difluorobiphenyl 98%
3,3'-Difluorobiphenyl

Precaución: for Research Use Only

Formula: C₁₂H₈F₂
MW: 190.19 g/mol
MDL Number: MFCD00039216
CAS Number: 396-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR