3-Ethoxy-5-(trifluoromethyl)picolinic acid 97%

Supplier: Ambeed
Warning

A906427-100MG A906427-1G A906427-250MG
77268-828EA 105.06 USD
77268-828 77268-830 77268-832
3-Ethoxy-5-(trifluoromethyl)picolinic acid 97%
3-Ethoxy-5-(trifluoromethyl)picolinic acid

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₃
MW: 235.16 g/mol
MDL Number: MFCD00205857
CAS Number: 257862-54-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR