(4-Benzoylphenyl)boronic acid 98%
Supplier: Ambeed
|
Warning
|
A906279-1G
A906279-250MG
A906279-5G
77294-216EA
23
USD
77294-216
77294-218
77294-220
(4-Benzoylphenyl)boronic acid 98%
(4-Benzoylphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₃H₁₁BO₃ |
MDL Number:
MFCD05664212 CAS Number: 268218-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...