4-n-Pentyloxyphenylacetylene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Eth-1-ynyl-4-(pentyloxy)benzene

A905979-1G A905979-5G
77244-744EA 10.3 USD
77244-744 77244-746
4-n-Pentyloxyphenylacetylene 98%
4-n-Pentyloxyphenylacetylene

Precaución: for Research Use Only

Formula: C₁₃H₁₆O
MW: 188.27 g/mol
MDL Number: MFCD00173877
CAS Number: 79887-16-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR