N-(2-Pyridyl)thiourea 98%

Supplier: Ambeed
Danger

A905406-10G A905406-1G A905406-25G A905406-5G
77281-870EA 22 USD
77281-870 77281-872 77281-874 77281-876
N-(2-Pyridyl)thiourea 98%
N-(2-Pyridyl)thiourea

Precaución: for Research Use Only

Formula: C₆H₇N₃S
MDL Number: MFCD00041227
CAS Number: 14294-11-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR