Tris(norbornadiene)(acetylacetonato)iridium(III) 98%

Supplier: Ambeed
Warning

A905289-100MG A905289-250MG
77268-824EA 47 USD
77268-824 77268-826
Tris(norbornadiene)(acetylacetonato)iridium(III) 98%
Tris(norbornadiene)(acetylacetonato)iridium(III)

Precaución: for Research Use Only

Formula: C₂₆H₃₁IrO₂
MDL Number: MFCD28167048
CAS Number: 41612-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR