4-Fluorostyrene 95% stabilized

Supplier: Ambeed
Danger

Synonyms: 1-Fluoro-4-vinylbenzene

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A904906-10G A904906-1G A904906-25G A904906-5G
77294-208EA 41.2 USD
77294-208 77294-210 77294-212 77294-214
4-Fluorostyrene 95% stabilized
4-Fluorostyrene

Precaución: for Research Use Only

Formula: C₈H₇F
MW: 122.14 g/mol
MDL Number: MFCD00000361
CAS Number: 405-99-2
UN: 1993
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR