4-(Cyclopropyl)benzyl bromide 97%

Supplier: Ambeed
Danger

Synonyms: 1-(Bromomethyl)-4-cyclopropylbenzene

A904465-1G A904465-250MG A904465-100MG
77268-814EA 370.8 USD
77268-814 77268-816 77749-718
4-(Cyclopropyl)benzyl bromide 97%
4-(Cyclopropyl)benzyl bromide

Precaución: for Research Use Only

Formula: C₁₀H₁₁Br
MW: 211.098 g/mol
MDL Number: MFCD12024391
CAS Number: 1150617-57-4
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR