2-Chloro-4'-fluoroacetanilide 95%

Supplier: Ambeed
Warning

A904070-1G A904070-5G
77244-732EA 10 USD
77244-732 77244-734
2-Chloro-4'-fluoroacetanilide 95%
2-Chloro-4'-fluoroacetanilide

Precaución: for Research Use Only

Formula: C₈H₇ClFNO
MDL Number: MFCD00015730
CAS Number: 351-04-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR