3-Ethylthiophene 95%

Supplier: Ambeed
Danger

A903739-25G A903739-5G
77281-866EA 157 USD
77281-866 77281-868
3-Ethylthiophene 95%
3-Ethylthiophene

Precaución: for Research Use Only

Formula: C₆H₈S
MDL Number: MFCD00070514
CAS Number: 1795-01-3
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR