5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinemanganese(III) chloride 98%

Supplier: Ambeed
Warning

A903064-100MG A903064-1G A903064-250MG
77256-526EA 31 USD
77256-526 77256-528 77256-530
5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinemanganese(III) chloride 98%
5,10,15,20-Tetrakis(4-methoxyphenyl)-21H,23H-porphinemanganese(III) chloride

Precaución: for Research Use Only

Formula: C₄₈H₃₆ClMnN₄O₄
MW: 823.21 g/mol
MDL Number: MFCD09038959
CAS Number: 62769-24-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR