3-Octen-2-one 97% (by GC)

Supplier: Ambeed

A901571-1G A901571-25G A901571-5G
77294-190EA 6.18 USD
77294-190 77294-192 77294-194
3-Octen-2-one 97% (by GC)
3-Octen-2-one

Precaución: for Research Use Only

Formula: C₈H₁₄O
MW: 126.20 g/mol
MDL Number: MFCD00015565
CAS Number: 1669-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR