3,5-Dibromo-2-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A901475-100G
A901475-10G
A901475-1G
A901475-25G
A901475-5G
77294-182EA
174
USD
77294-182
77294-184
77294-186
77294-188
77749-711
3,5-Dibromo-2-fluoropyridine 98%
3,5-Dibromo-2-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₂Br₂FN |
MDL Number:
MFCD07368619 CAS Number: 473596-07-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...