2-(Chloromethyl)-5-methyl-1,3,4-oxadiazole 95%

Supplier: Ambeed
Danger

A901397-100MG A901397-1G A901397-250MG A901397-5G
77244-722EA 18.54 USD
77244-722 77244-724 77244-726 77244-728
2-(Chloromethyl)-5-methyl-1,3,4-oxadiazole 95%
2-(Chloromethyl)-5-methyl-1,3,4-oxadiazole

Precaución: for Research Use Only

Formula: C₄H₅ClN₂O
MW: 132.54 g/mol
MDL Number: MFCD06655111
CAS Number: 3914-42-9
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR