4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Fluorobenzoyl)-1,1,1-trifluoroacetone

A901236-250MG A901236-5G
77294-178EA 26.78 USD
77294-178 77294-180
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione 95%
4,4,4-Trifluoro-1-(4-fluorophenyl)butane-1,3-dione

Precaución: for Research Use Only

Formula: C₁₀H₆F₄O₂
MW: 234.15 g/mol
MDL Number: MFCD03419766
CAS Number: 582-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR