1-(4-Isocyanatophenyl)ethanone 98%

Supplier: Ambeed
Danger

Synonyms: Isocyanic acid 4-Acetylphenyl ester, 4-Acetylphenylisocyanate, 4-Acetylphenyl Isocyanate

A899249-1G A899249-5G
77268-806EA 30 USD
77268-806 77268-808
1-(4-Isocyanatophenyl)ethanone 98%
1-(4-Isocyanatophenyl)ethanone

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MDL Number: MFCD00013879
CAS Number: 49647-20-3
UN: 2206
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR