1-Bromo-2-(methoxymethyl)benzene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromobenzyl methyl ether

A898571-1G A898571-250MG A898571-25G
77244-706EA 19.57 USD
77244-706 77244-708 77244-710
1-Bromo-2-(methoxymethyl)benzene 98%
1-Bromo-2-(methoxymethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₉BrO
MW: 201.0605 g/mol
MDL Number: MFCD03790887
CAS Number: 52711-30-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR