2-Aminoethyl methacrylate hydrochloride 90% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Aminoethyl 2-methylprop-2-enoate hydrochloride
Stabilization:
~500 ppm Stabilised with phenothiazine
A897686-1G
A897686-250MG
A897686-25G
A897686-5G
77268-798EA
11
USD
77268-798
77268-800
77268-802
77268-804
2-Aminoethyl methacrylate hydrochloride 90% stabilised
2-Aminoethyl methacrylate hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₁₂ClNO₂ MW: 165.617 g/mol |
MDL Number:
MFCD00078260 CAS Number: 2420-94-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...