Tetrahydro-2H-thiopyran-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 4-Carboxytetrahydro-2H-thiopyran, 4-Carboxythiane, Tetrahydrothiopyran-4-carboxylic Acid

A897572-1G A897572-5G
77268-794EA 20 USD
77268-794 77268-796
Tetrahydro-2H-thiopyran-4-carboxylic acid 98%
Tetrahydro-2H-thiopyran-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₁₀O₂S
MDL Number: MFCD09879921
CAS Number: 89489-53-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR