SRT-3025 hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5-(3-methoxypropyl)-2-phenyl-N-(2-(6-(pyrrolidin-1-ylmethyl)thiazolo[5,4-b]pyridin-2-yl)phenyl)thiazole-4-carboxamide hydrochloride, SRT3025 HCl, Sirtuin modulator 1

A896798-1MG A896798-5MG
77268-784EA 17.51 USD
77268-784 77268-786
SRT-3025 hydrochloride 98%
SRT-3025 hydrochloride

Precaución: for Research Use Only

Formula: C₃₁H₃₂ClN₅O₂S₂
MW: 606.2 g/mol
MDL Number: MFCD29924722
CAS Number: 2070015-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR