1-((2,3-Dihydrobenzo[b][1,4]dioxin-2-yl)methyl)-3-hydroxythieno[3,2-d]pyrimidine-2,4(1H,3H)-dione 98%

Supplier: Ambeed
Warning

Synonyms: FEN1-IN-1

A896411-10MG A896411-1MG A896411-5MG
77268-778EA 45.32 USD
77268-778 77268-780 77268-782
1-((2,3-Dihydrobenzo[b][1,4]dioxin-2-yl)methyl)-3-hydroxythieno[3,2-d]pyrimidine-2,4(1H,3H)-dione 98%
1-((2,3-Dihydrobenzo[b][1,4]dioxin-2-yl)methyl)-3-hydroxythieno[3,2-d]pyrimidine-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂O₅S
MW: 332.33 g/mol
MDL Number: MFCD32263021
CAS Number: 824983-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR