(R)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine 97%

Supplier: Ambeed
Warning

A895748-1G A895748-250MG A895748-5G
77256-508EA 41.2 USD
77256-508 77256-510 77256-512
(R)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine 97%
(R)-1-(3-Ethoxy-4-methoxyphenyl)-2-(methylsulfonyl)ethanamine

Precaución: for Research Use Only

Formula: C₁₂H₁₉NO₄S
MW: 273.34 g/mol
MDL Number: MFCD22643957
CAS Number: 608142-27-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR