6-Indoleboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: Indole-6-boronic acid

A895029-1G A895029-250MG A895029-5G
77281-858EA 12.36 USD
77281-858 77281-860 77281-862 77281-864
6-Indoleboronic acid 97%
6-Indoleboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BNO₂
MW: 160.97 g/mol
MDL Number: MFCD03095176
CAS Number: 147621-18-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR