(R)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%

Supplier: Ambeed
Warning

A894906-1G A894906-5G
77244-692EA 35.02 USD
77244-692 77618-504
(R)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid 97%
(R)-3-(4-(tert-Butoxy)phenyl)-2-((tert-butoxycarbonyl)amino)propanoic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₇NO₅
MW: 337.41 g/mol
MDL Number: MFCD01861337
CAS Number: 507276-74-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR