UNBS-5162 98%

Supplier: Ambeed
Warning

Synonyms: UNBS5162, 1-(2-(2-(Dimethylamino)ethyl)-1,3-dioxo-2,3-dihydro-1H-benzo[de]isoquinolin-5-yl)urea

A893890-10MG A893890-1MG A893890-25MG A893890-50MG
77256-500EA 224.54 USD
77256-500 77256-502 77256-504 77256-506
UNBS-5162 98%
UNBS-5162

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₄O₃
MW: 326.35 g/mol
MDL Number: MFCD22380608
CAS Number: 956590-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR