2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide 95%

Supplier: Ambeed
Warning

Synonyms: 4-[(Bromoacetyl)amino]benzotrifluoride, N-(Bromoacetyl)-4-(trifluoromethyl)aniline

A893639-1G A893639-250MG A893639-5G
77268-770EA 59 USD
77268-770 77268-772 77268-774
2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide 95%
2-Bromo-N-(4-(trifluoromethyl)phenyl)acetamide

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO
MW: 282.06 g/mol
MDL Number: MFCD00662747
CAS Number: 3823-19-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR