5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Acetyl-3-chloro-10,11-dihydro-5H-dibenz[b,f]azepine, 5-Acetyl-3-chloro-2,2'-iminodibenzyl

A893405-100G A893405-1G A893405-25G A893405-5G
77281-850EA 103 USD
77281-850 77281-852 77281-854 77281-856
5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine 98%
5-Acetyl-3-chloro-10,11-dihydrodibenzo[b,f]azepine

Precaución: for Research Use Only

Formula: C₁₆H₁₄ClNO
MW: 271.74 g/mol
MDL Number: MFCD01632170
CAS Number: 25961-11-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR