1-Bromo-4-isobutylbenzene 98%

Supplier: Ambeed
Warning

A892840-100G A892840-10G A892840-1G A892840-25G A892840-5G
77244-682EA 285.31 USD
77244-682 77244-684 77244-686 77244-688 77244-690
1-Bromo-4-isobutylbenzene 98%
1-Bromo-4-isobutylbenzene

Precaución: for Research Use Only

Formula: C₁₀H₁₃Br
MDL Number: MFCD03425886
CAS Number: 2051-99-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR