Ethynyl-p-tolyl sulfone 98%

Supplier: Ambeed
Warning

Synonyms: Tosylacetylene, Ethynyl p-Tolyl sulphone, p-Toluenesulphonylacetylene

A892225-1G A892225-250MG
77268-766EA 91.67 USD
77268-766 77268-768
Ethynyl-p-tolyl sulfone 98%
Ethynyl-p-tolyl sulfone

Precaución: for Research Use Only

Formula: C₉H₈O₂S
MW: 180.22 g/mol
MDL Number: MFCD00191647
CAS Number: 13894-21-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR