Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside 98%

Supplier: Ambeed
Warning

Synonyms: (2R,3R,4S,5S,6R)-2-(Acetoxymethyl)-6-(phenylthio)tetrahydro-2H-pyran-3,4,5-triyl triacetate

A891683-1G A891683-250MG A891683-5G
77244-676EA 52 USD
77244-676 77244-678 77244-680
Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside 98%
Phenyl 2,3,4,6-Tetra-O-acetyl-1-thio-alpha-D-mannopyranoside

Precaución: for Research Use Only

Formula: C₂₀H₂₄O₉S
MW: 440.46 g/mol
MDL Number: MFCD01862644
CAS Number: 108032-93-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR