N1,N2-Dimethylphthalamide 97%

Supplier: Ambeed
Warning

A891610-100MG A891610-1G A891610-250MG
77244-670EA 46.35 USD
77244-670 77244-672 77244-674
N1,N2-Dimethylphthalamide 97%
N1,N2-Dimethylphthalamide

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₂
MW: 192.21 g/mol
MDL Number: MFCD00086741
CAS Number: 19532-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR