4-(Dimethylamino)-3-nitrobenzenecarbaldehyde 95+%

Supplier: Ambeed
Warning

A891139-1G A891139-250MG A891139-5G
77294-154EA 15 USD
77294-154 77294-156 77294-158
4-(Dimethylamino)-3-nitrobenzenecarbaldehyde 95+%
4-(Dimethylamino)-3-nitrobenzenecarbaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀N₂O₃
MDL Number: MFCD00480831
CAS Number: 59935-39-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR