2-Chloroethanesulfonic acid sodium salt hydrate 95%
Supplier: Ambeed
|
Warning
|
A889564-1000G
A889564-100G
A889564-25G
A889564-500G
A889564-5G
77268-750EA
35
USD
77268-750
77268-752
77268-754
77268-756
77268-758
2-Chloroethanesulfonic acid sodium salt hydrate 95%
2-Chloroethanesulfonic acid sodium salt hydrate
Precaución: for Research Use Only
Formula:
C₂H₄ClNaO₃S.H₂O |
MDL Number:
MFCD00007531 CAS Number: 1089719-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...