6,6'-Diamino-2,2'-bipyridyl 98%

Supplier: Ambeed
Warning

Synonyms: [2,2'-Bipyridine]-6,6'-diamine, 2,2'-Bipyridine-6,6'-diamine

A889318-100MG A889318-250MG A889318-1G
77256-490EA 61.8 USD
77256-490 77256-492 77308-490
6,6'-Diamino-2,2'-bipyridyl 98%
6,6'-Diamino-2,2'-bipyridyl

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₄
MW: 186.22 g/mol
MDL Number: MFCD04117918
CAS Number: 93127-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR