4-Bromothioanisole 99%

Supplier: Ambeed
Danger

A887360-100G A887360-10G A887360-25G A887360-5G
77294-136EA 72 USD
77294-136 77294-138 77294-140 77294-142 77294-144
4-Bromothioanisole 99%
4-Bromothioanisole

Precaución: for Research Use Only

Formula: C₇H₇BrS
MDL Number: MFCD00000102
CAS Number: 104-95-0
UN: 3335
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR