(S)-1-(2-(Difluoromethyl)pyridin-4-yl)-4-fluoro-1-(3-(pyrimidin-5-yl)phenyl)-1H-isoindol-3-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: AZD3839

A886914-1MG A886914-5MG
77268-734EA 40.17 USD
77268-734 77268-736
(S)-1-(2-(Difluoromethyl)pyridin-4-yl)-4-fluoro-1-(3-(pyrimidin-5-yl)phenyl)-1H-isoindol-3-amine ≥98%
(S)-1-(2-(Difluoromethyl)pyridin-4-yl)-4-fluoro-1-(3-(pyrimidin-5-yl)phenyl)-1H-isoindol-3-amine

Precaución: for Research Use Only

Formula: C₂₄H₁₆F₃N₅
MW: 431.41 g/mol
MDL Number: MFCD25976574
CAS Number: 1227163-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR