2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine xhydrochloride 95%

Supplier: Ambeed
Warning

A886285-1G A886285-250MG A886285-5G
77268-728EA 139.05 USD
77268-728 77268-730 77268-732
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine xhydrochloride 95%
2,3-Dihydro-1H-pyrrolo[3,4-c]pyridine xhydrochloride

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MW: 156.61 g/mol
CAS Number: 1255306-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR