Methyl-4-fluorobenzo[b]thiophene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 4-fluorobenzo[b]thiophene-2-carboxylate

A886183-100MG A886183-1G A886183-250MG A886183-5G
77294-128EA 5 USD
77294-128 77294-130 77294-132 77294-134
Methyl-4-fluorobenzo[b]thiophene-2-carboxylate 98%
Methyl 4-fluorobenzo[b]thiophene-2-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₇FO₂S
MDL Number: MFCD06200964
CAS Number: 220180-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR