4-Methyl-1,4'-bipiperidine 96%

Supplier: Ambeed
Warning

A886128-100MG A886128-1G A886128-250MG A886128-5G
77294-120EA 11.33 USD
77294-120 77294-122 77294-124 77294-126
4-Methyl-1,4'-bipiperidine 96%
4-Methyl-1,4'-bipiperidine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂
MW: 182.30 g/mol
MDL Number: MFCD03274731
CAS Number: 116797-02-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR