3-(Phenylsulfanyl)propanoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(Phenylthio)propanoic acid

A886026-100MG A886026-1G A886026-250MG
77281-844EA 63.86 USD
77281-844 77281-846 77281-848
3-(Phenylsulfanyl)propanoic acid 95%
3-(Phenylsulfanyl)propanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₂S
MW: 182.24 g/mol
MDL Number: MFCD00215509
CAS Number: 5219-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR