2-(5-Bromo-2-thienyl)ethylamine 97%

Supplier: Ambeed
Warning

A884866-100MG A884866-1G A884866-250MG A884866-5G
77244-656EA 56.65 USD
77244-656 77244-658 77244-660 77244-662
2-(5-Bromo-2-thienyl)ethylamine 97%
2-(5-Bromo-2-thienyl)ethylamine

Precaución: for Research Use Only

Formula: C₆H₈BrNS
MW: 206.10 g/mol
MDL Number: MFCD04114538
CAS Number: 86423-64-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR