(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-((S)-2-oxopyrrolidin-3-yl)propanoate 97%

Supplier: Ambeed
Warning

Synonyms: Paxlovid Intermediate 3

A884242-100G A884242-1G A884242-250MG A884242-25G A884242-5G
77244-644EA 144 USD
77244-644 77244-646 77244-648 77244-650 77244-652
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-((S)-2-oxopyrrolidin-3-yl)propanoate 97%
(S)-Methyl 2-((tert-butoxycarbonyl)amino)-3-((S)-2-oxopyrrolidin-3-yl)propanoate

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₂O₅
MDL Number: MFCD28138267
CAS Number: 328086-60-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR