tert-Butyl (6-iodohexyl)carbamate 97%

Supplier: Ambeed
Warning

A883059-100MG A883059-1G A883059-250MG A883059-5G
77256-478EA 43 USD
77256-478 77256-480 77256-482 77256-484
tert-Butyl (6-iodohexyl)carbamate 97%
tert-Butyl (6-iodohexyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂INO₂
MDL Number: MFCD09951827
CAS Number: 172846-36-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR