BMS-986205 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-N-(4-Chlorophenyl)-2-(cis-4-(6-fluoroquinolin-4-yl)cyclohexyl)propanamide, Linrodostat, (R)-N-(4-chlorophenyl)-2-((1s,4S)-4-(6-fluoroquinolin-4-yl)cyclohexyl)propenamide

A883044-1MG A883044-25MG A883044-50MG A883044-5MG A883044-10MG
77268-714EA 26.78 USD
77268-714 77268-716 77268-718 77268-720 77749-686
BMS-986205 98+%
BMS-986205

Precaución: for Research Use Only

Formula: C₂₄H₂₄ClFN₂O
MW: 410.92 g/mol
MDL Number: MFCD31729367
CAS Number: 1923833-60-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR